Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp978 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025677.1 | NP_001020848.1 | 4 | 925 | ||
XM_006228178.2 | XP_006228240.1 | 5 | 1075 | |||
GenBank mRNA | BC097363.1 | - | 4 | 925 | 7442984 |