Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lamtor1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199102.1 | NP_954533.1 | 3, 4 | 317 | ||
GenBank mRNA | BC061783.1 | - | 3, 4 | 317 | 5623933 | |
FQ214531.1 | - | 3, 4 | 321 | |||
FQ219988.1 | - | 3, 4 | 330 | |||
FQ220581.1 | - | 3, 4 | 313 | |||
FQ220693.1 | - | 3, 4 | 315 |
Gene Symbol: LOC100361543 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002730236.3 | XP_002730282.1 | 1 | 294 |