Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107542.2 | NP_001101012.2 | 27, 28 | 4249 | ||
XM_006230044.2 | XP_006230106.1 | 29, 30 | 4193 | |||
XM_006230045.2 | XP_006230107.1 | 28, 29 | 4373 | |||
XM_006230046.2 | XP_006230108.1 | 28, 29 | 4133 | |||
GenBank mRNA | CF109520.1 | - | 5, 6 | 456 | ||
CO571839.1 | - | 6, 7 | 616 |