Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107542.2 | NP_001101012.2 | 23, 24 | 3825 | ||
XM_006230044.2 | XP_006230106.1 | 25, 26 | 3769 | |||
XM_006230045.2 | XP_006230107.1 | 24, 25 | 3949 | |||
XM_006230046.2 | XP_006230108.1 | 24, 25 | 3709 | |||
GenBank mRNA | CF109520.1 | - | 1, 2 | 32 | ||
CO571839.1 | - | 2, 3 | 192 |