Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcun1d3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024886.1 | NP_001020057.1 | 3 | 870 | ||
XM_008759750.1 | XP_008757972.1 | 3 | 796 | |||
GenBank mRNA | BC097462.1 | - | 3 | 870 | 7457575 |