Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107637.1 | NP_001101107.1 | 2 | 409 | ||
XM_006256570.2 | XP_006256632.1 | 2 | 289 | |||
XM_006256571.2 | XP_006256633.1 | 2 | 290 |