Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RAPGEF2 SIRNA Assays ›
          Gene Symbol:
          Rapgef2
          Gene Name:
          Rap guanine nucleotide exchange factor (GEF) 2
          Gene Aliases:
          PDZ-GEF1, RA-GEF-1, nRap GEP
          Chromosome Location:
          Chr.2: 177836191 - 178057397 on Build Rnor_6.0
          UniGene ID:
          Rn.23121
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID s160418

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rapgef2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001107684.1 NP_001101154.1 1 99
          XM_006232511.2 XP_006232573.2 2 1700
          XM_008761092.1 XP_008759314.1 6 1070
          XM_008761093.1 XP_008759315.1 6 1071
          XM_008761094.1 XP_008759316.1 6 1070
          XM_008761095.1 XP_008759317.1 6 744
          XM_008761096.1 XP_008759318.1 4 1359
          XM_008761097.1 XP_008759319.1 3 1251
          XM_008761098.1 XP_008759320.1 3 2543

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          blood vessel development
          neuron migration
          G-protein coupled receptor signaling pathway
          neuropeptide signaling pathway
          small GTPase mediated signal transduction
          negative regulation of cell proliferation
          positive regulation of neuron projection development
          cAMP-mediated signaling
          ventricular system development
          forebrain neuron development
          microvillus assembly
          neuron projection development
          brain-derived neurotrophic factor receptor signaling pathway
          positive regulation of protein binding
          Rap protein signal transduction
          nerve growth factor signaling pathway
          positive regulation of GTPase activity
          positive regulation of cAMP-mediated signaling
          positive regulation of protein kinase activity
          negative regulation of melanin biosynthetic process
          regulation of synaptic plasticity
          negative regulation of dendrite morphogenesis
          regulation of small GTPase mediated signal transduction
          establishment of endothelial barrier
          positive regulation of ERK1 and ERK2 cascade
          cellular response to cAMP
          cellular response to cGMP
          adenylate cyclase-activating adrenergic receptor signaling pathway
          protein localization to plasma membrane
          establishment of endothelial intestinal barrier
          regulation of cell junction assembly
          cellular response to nerve growth factor stimulus
          positive regulation of cAMP-dependent protein kinase activity
          positive regulation of dendritic cell apoptotic process
          positive regulation of vasculogenesis
          positive regulation of neuron migration
          guanyl-nucleotide exchange factor activity
          Ras guanyl-nucleotide exchange factor activity
          protein binding
          Rap guanyl-nucleotide exchange factor activity
          protein kinase binding
          PDZ domain binding
          cAMP binding
          cGMP binding
          beta-1 adrenergic receptor binding
          WW domain binding
          phosphatidic acid binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b7b59cd5b-z8v7b:80/100.66.130.62:80.
          git-commit: 87deb889a5bb48f859ebd9970e87be562fd9396c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0