Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrpl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007696.1 | NP_001007697.1 | 5, 6 | 601 | ||
GenBank mRNA | BC082085.1 | - | 5, 6 | 601 | 7133689 |