Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DGKD SIRNA Assays ›
          Gene Symbol:
          DGKD
          Gene Name:
          diacylglycerol kinase delta
          Gene Aliases:
          DGKdelta, dgkd-2
          Chromosome Location:
          Chr.2: 233354507 - 233472098 on Build GRCh38
          UniGene ID:
          Hs.471675
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s16211

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DGKD
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_003648.2 NP_003639.2 17, 18 2210
          NM_152879.2 NP_690618.2 18, 19 2275
          XM_005246107.3 XP_005246164.1 16, 17 1942
          XM_005246108.1 XP_005246165.1 16, 17 2096
          XM_005246110.3 XP_005246167.1 7, 8 1947
          XM_011512028.2 XP_011510330.1 18, 19 3734
          XM_011512029.2 XP_011510331.1 18, 19 3735
          XM_011512030.2 XP_011510332.1 18, 19 2331
          XM_011512031.2 XP_011510333.1 18, 19 2205
          XM_011512032.2 XP_011510334.1 18, 19 2189
          XM_011512033.2 XP_011510335.1 18, 19 3736
          XM_011512035.1 XP_011510337.1 16, 17 2076
          XM_011512037.2 XP_011510339.1 16, 17 2180
          XM_011512039.2 XP_011510341.1 18, 19 3736
          XM_011512040.2 XP_011510342.1 16, 17 2112
          XM_017005138.1 XP_016860627.1 18, 19 2613
          XM_017005139.1 XP_016860628.1 16, 17 2105
          XM_017005140.1 XP_016860629.1 18, 19 2207
          GenBank mRNA AB078966.1 - 18, 19 2275
          AK307426.1 - 6, 7 833
          AK308846.1 - 16, 17 2096
          AK311044.1 - 16, 17 2029
          D63479.3 - 18, 19 2216
          D73409.1 - 17, 18 2211

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          endocytosis
          signal transduction
          epidermal growth factor receptor signaling pathway
          protein kinase C-activating G-protein coupled receptor signaling pathway
          multicellular organism development
          response to organic substance
          protein transport
          cell growth
          phosphorylation
          second-messenger-mediated signaling
          platelet activation
          diacylglycerol metabolic process
          protein homooligomerization
          diacylglycerol kinase activity
          protein binding
          ATP binding
          diacylglycerol binding
          protein homodimerization activity
          metal ion binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix