Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EPB4.1 SIRNA Assays ›
Gene Symbol:
Epb4.1
Gene Name:
erythrocyte membrane protein band 4.1
Gene Aliases:
RGD1564762
Chromosome Location:
Chr.5: 150081727 - 150243186 on Build Rnor_6.0
UniGene ID:
Rn.22747
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s162150
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Epb4.1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
XM_006239085.2
XP_006239147.1
10
1711
XM_006239086.2
XP_006239148.1
10
1711
XM_006239087.2
XP_006239149.1
10
3663
XM_006239088.2
XP_006239150.1
10
1711
XM_006239089.2
XP_006239151.1
10
1711
XM_006239090.2
XP_006239152.1
10
1711
XM_006239091.2
XP_006239153.1
10
1711
XM_006239092.2
XP_006239154.1
10
1712
XM_006239093.2
XP_006239155.1
10
1547
XM_006239094.2
XP_006239156.1
10
1711
XM_006239095.1
XP_006239157.1
10
1570
XM_006239096.2
XP_006239158.1
10
1709
XM_006239097.2
XP_006239159.1
10
1709
XM_006239098.2
XP_006239160.1
10
1709
XM_006239099.2
XP_006239161.1
10
1709
XM_006239100.2
XP_006239162.1
10
1708
XM_006239101.2
XP_006239163.1
10
1710
XM_006239103.2
XP_006239165.1
10
1708
XM_006239104.2
XP_006239166.1
10
1709
XM_006239105.2
XP_006239167.1
10
1711
XM_008764184.1
XP_008762406.1
10
1708
XM_008764185.1
XP_008762407.1
10
1708
XM_232771.8
XP_232771.6
10
1547
GenBank mRNA
FQ233548.1
-
10
1506
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
cortical actin cytoskeleton organization
positive regulation of protein binding
actin binding
structural molecule activity
protein binding
1-phosphatidylinositol binding
spectrin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.32.0-2025.07.28-1.0