Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mob3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107960.1 | NP_001101430.1 | 2 | 291 | ||
XM_006238657.2 | XP_006238719.1 | 1 | 1207 | |||
XM_006238658.2 | XP_006238720.1 | 2 | 712 | |||
XM_006238659.2 | XP_006238721.1 | 2 | 2209 | |||
XM_008763977.1 | XP_008762199.1 | 1 | 1207 |