Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mroh4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765606.1 | XP_008763828.1 | 18 | 2234 | ||
XM_008765607.1 | XP_008763829.1 | 18 | 2505 | |||
XM_008765608.1 | XP_008763830.1 | 18 | 3621 | |||
XM_008765609.1 | XP_008763831.1 | 18 | 3637 | |||
XM_008765610.1 | XP_008763832.1 | 18 | 2522 | |||
XM_008765611.1 | XP_008763833.1 | 18 | 2250 | |||
XM_008765612.1 | XP_008763834.1 | 19 | 2407 | |||
XM_008765613.1 | XP_008763835.1 | 17 | 2289 | |||
XM_008765614.1 | XP_008763836.1 | 18 | 2234 | |||
XM_008765615.1 | XP_008763837.1 | 18 | 2102 | |||
XM_008765616.1 | XP_008763838.1 | 17 | 2133 | |||
XM_008765617.1 | XP_008763839.1 | 18 | 2234 | |||
XM_008765618.1 | XP_008763840.1 | 18 | 2234 |