Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013176.1 | NP_001013194.1 | 3 | 435 | ||
XM_006243035.2 | XP_006243097.1 | 4 | 993 | |||
XM_006243036.2 | XP_006243098.1 | 3 | 393 | |||
XM_006243038.2 | XP_006243100.1 | 3 | 290 | |||
XM_008766262.1 | XP_008764484.1 | 3 | 427 | |||
GenBank mRNA | AY960132.1 | - | 2 | 240 | ||
BC088755.1 | - | 3 | 435 | 7307514 |