Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtfmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009697.1 | NP_001009697.1 | 9 | 1117 | ||
GenBank mRNA | BC088470.1 | - | 9 | 1117 | 7128152 | |
FQ226710.1 | - | 5 | 444 |