Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012130.1 | NP_001012130.1 | 1 | 158 | ||
GenBank mRNA | BC083568.1 | - | 1 | 158 | 7111073 |