Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Farp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108233.1 | NP_001101703.1 | 22, 23 | 2617 | ||
XM_006245529.2 | XP_006245591.1 | 23, 24 | 2645 | |||
XM_006245530.2 | XP_006245592.1 | 24, 25 | 2721 | |||
XM_006245532.2 | XP_006245594.1 | 22, 23 | 2624 | |||
XM_008767375.1 | XP_008765597.1 | 23, 24 | 2578 |