Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025124.1 | NP_001020295.1 | 432 | |||
GenBank mRNA | BC091175.1 | - | 432 | 7321868 | ||
FQ216165.1 | - | 430 | ||||
FQ219051.1 | - | 325 | ||||
FQ230577.1 | - | 428 |
Gene Symbol: LOC103690022 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767472.1 | XP_008765694.1 | 3 | 330 | ||
XM_008767473.1 | XP_008765695.1 | 3 | 330 | |||
XM_008767474.1 | XP_008765696.1 | 3 | 330 | |||
XM_008767475.1 | XP_008765697.1 | 3 | 330 | |||
XM_008767476.1 | XP_008765698.1 | 5 | 470 |