Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo18a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172137.1 | NP_001165608.1 | 37 | 5666 | ||
GenBank mRNA | BC105827.1 | - | 2 | 165 |