Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ANK3 SIRNA Assays ›
          Gene Symbol:
          Ank3
          Gene Name:
          ankyrin 3, node of Ranvier (ankyrin G)
          Gene Aliases:
          ANK-3
          Chromosome Location:
          Chr.20: 19948767 - 20480628 on Build Rnor_6.0
          UniGene ID:
          Rn.211093
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID s168097

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ank3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001033984.1 NP_001029156.1 41 7203
          NM_031805.1 NP_113993.1 37 4849
          GenBank mRNA AF065149.1 - 3 231
          AF069525.1 - 37 4707
          AF102552.1 - 41 7203
          AJ428573.1 - 16 2032
          AJ812019.1 - 16 2392
          AJ812020.1 - 16 2392
          AJ812021.1 - 16 2392
          AJ812022.1 - 16 2392
          AJ812023.1 - 37 4841
          AJ812024.1 - 37 4841
          BC101885.1 - 16 2073

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_170687, NM_170729, NM_170728, NM_170690, NM_009670, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic cytokinesis
          plasma membrane organization
          signal transduction
          axonogenesis
          axon guidance
          neuromuscular junction development
          positive regulation of gene expression
          positive regulation of cell communication by electrical coupling
          positive regulation of sodium ion transport
          magnesium ion homeostasis
          neuronal action potential
          positive regulation of homotypic cell-cell adhesion
          Golgi to plasma membrane protein transport
          regulation of potassium ion transport
          establishment of protein localization
          positive regulation of membrane potential
          synapse organization
          cellular response to magnesium ion
          membrane assembly
          protein localization to plasma membrane
          maintenance of protein location in plasma membrane
          protein targeting to plasma membrane
          positive regulation of protein targeting to membrane
          positive regulation of membrane depolarization during cardiac muscle cell action potential
          negative regulation of delayed rectifier potassium channel activity
          positive regulation of sodium ion transmembrane transporter activity
          positive regulation of cation channel activity
          structural constituent of cytoskeleton
          protein binding
          cytoskeletal protein binding
          spectrin binding
          protein binding, bridging
          ion channel binding
          cadherin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0