Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012184.1 | NP_001012184.1 | 8, 9 | 1243 | ||
XM_006234234.2 | XP_006234296.1 | 8, 9 | 1000 | |||
XM_006234235.2 | XP_006234297.1 | 8, 9 | 1293 | |||
XM_006234236.2 | XP_006234298.1 | 8, 9 | 1092 | |||
XM_006234237.2 | XP_006234299.1 | 8, 9 | 1852 | |||
XM_006234238.2 | XP_006234300.1 | 8, 9 | 1000 | |||
XM_006234239.2 | XP_006234301.1 | 9, 10 | 1490 | |||
XM_006234241.2 | XP_006234303.1 | 8, 9 | 1090 | |||
XM_008761860.1 | XP_008760082.1 | 8, 9 | 1581 | |||
GenBank mRNA | BC087094.1 | - | 8, 9 | 1243 | 7191334 |