Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scmh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109669.1 | NP_001103139.1 | 12 | 1695 | ||
XM_008764027.1 | XP_008762249.1 | 12 | 2179 | |||
XM_008764028.1 | XP_008762250.1 | 12 | 2179 | |||
XM_008764029.1 | XP_008762251.1 | 11 | 2039 | |||
XM_008764030.1 | XP_008762252.1 | 11 | 4256 | |||
XM_008764031.1 | XP_008762253.1 | 12 | 1786 | |||
XM_008764032.1 | XP_008762254.1 | 10 | 1543 | |||
GenBank mRNA | BC131847.1 | - | 12 | 1758 | 7100459 |