Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SPHK1 SIRNA Assays ›
          Gene Symbol:
          SPHK1
          Gene Name:
          sphingosine kinase 1
          Gene Aliases:
          SPHK
          Chromosome Location:
          Chr.17: 76382977 - 76387860 on Build GRCh38
          UniGene ID:
          Hs.68061
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s16958
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SPHK1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001142601.1 NP_001136073.1 6 1046
          NM_001142602.1 NP_001136074.1 5 1091
          NM_021972.3 NP_068807.2 6 1088
          NM_182965.2 NP_892010.2 6 1409
          XM_005257766.2 XP_005257823.1 6 1401
          GenBank mRNA AF200328.1 - 5 672
          AF238083.1 - 6 990
          AF266756.1 - 5 923
          AJ245504.1 - 4 645
          AK022402.1 - 6 1046
          AK023393.1 - 5 1003
          AK095578.1 - 5 1731
          AK122781.1 - 8 1463
          AK225250.1 - 6 1009
          AK225258.1 - 6 875
          AK292294.1 - 6 1133
          AK292614.1 - 5 1007
          BC004112.2 - 2 363
          BC008040.2 - 6 983 3831657
          BC009419.1 - 6 1076 3506924
          BC014439.1 - 6 1346 4871343
          BC030553.1 - 6 1368 5213270

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00184211_m1

          Gene Ontology Categories:

          Function(s) Process(es)

          blood vessel development
          positive regulation of neurotransmitter secretion
          sphingosine-1-phosphate signaling pathway
          protein folding
          sphingosine metabolic process
          inflammatory response
          signal transduction
          brain development
          female pregnancy
          cellular response to starvation
          positive regulation of peptidyl-threonine phosphorylation
          regulation of tumor necrosis factor-mediated signaling pathway
          positive regulation of neuron projection development
          response to amine
          cyclooxygenase pathway
          calcium-mediated signaling
          sphingolipid biosynthetic process
          positive regulation of cell growth
          positive regulation of cell migration
          positive regulation of protein ubiquitination
          response to magnesium ion
          response to progesterone
          regulation of interleukin-1 beta production
          response to ATP
          intracellular signal transduction
          positive regulation of NF-kappaB import into nucleus
          negative regulation of apoptotic process
          positive regulation of angiogenesis
          positive regulation of mitotic cell cycle
          positive regulation of smooth muscle contraction
          sphingosine biosynthetic process
          sphingoid catabolic process
          lipid phosphorylation
          positive regulation of fibroblast proliferation
          positive regulation of NF-kappaB transcription factor activity
          cellular response to hydrogen peroxide
          response to interleukin-1
          cellular response to growth factor stimulus
          magnesium ion binding
          DNA binding
          protein binding
          calmodulin binding
          ATP binding
          sphinganine kinase activity
          D-erythro-sphingosine kinase activity
          sphingosine-1-phosphate receptor activity
          protein phosphatase 2A binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0