Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034002.2 | NP_001029174.2 | 3 | 530 | ||
XM_006242488.2 | XP_006242550.1 | 3 | 1177 | |||
XM_006242489.2 | XP_006242551.1 | 3 | 1175 | |||
XM_006242490.2 | XP_006242552.1 | 3 | 1176 | |||
XM_006242491.2 | XP_006242553.1 | 3 | 1174 | |||
XM_006242492.2 | XP_006242554.1 | 3 | 1176 | |||
XM_006242493.2 | XP_006242555.1 | 3 | 1174 | |||
XM_008765893.1 | XP_008764115.1 | 3 | 1177 | |||
XM_008765894.1 | XP_008764116.1 | 3 | 1175 | |||
GenBank mRNA | DQ186896.2 | - | 3 | 530 | ||
DQ186897.2 | - | 3 | 530 | |||
DQ186898.2 | - | 3 | 530 | |||
DQ376007.1 | - | 3 | 530 |