Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Speg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108802.1 | NP_001102272.1 | 9 | 2991 | ||
NM_012905.2 | NP_037037.1 | 4 | 479 | |||
XM_006245252.2 | XP_006245314.1 | 11 | 2953 | |||
XM_006245255.2 | XP_006245317.1 | 4 | 2749 | |||
XM_006245256.2 | XP_006245318.1 | 4 | 778 | |||
XM_008767267.1 | XP_008765489.1 | 9 | 3053 | |||
XM_008767268.1 | XP_008765490.1 | 9 | 2830 | |||
XM_008767269.1 | XP_008765491.1 | 10 | 3447 | |||
XM_008767270.1 | XP_008765492.1 | 9 | 3006 | |||
XM_008767271.1 | XP_008765493.1 | 9 | 3005 | |||
GenBank mRNA | BC087690.1 | - | 1 | 117 | ||
EV763588.1 | - | 4 | 479 | |||
U57097.1 | - | 4 | 455 |