Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp609 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173371.1 | NP_001166842.1 | 1 | 177 | ||
XM_008766321.1 | XP_008764543.1 | 1 | 3066 |