Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eef1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004223.2 | NP_001004223.1 | 6, 7 | 857 | ||
GenBank mRNA | BC079398.1 | - | 7, 8 | 866 | 7112627 | |
BC098895.1 | - | 6, 7 | 857 | 7367246 | ||
FQ212596.1 | - | 7, 8 | 875 | |||
FQ213179.1 | - | 7, 8 | 874 | |||
FQ214210.1 | - | 7, 8 | 872 | |||
FQ214443.1 | - | 7, 8 | 875 | |||
FQ215853.1 | - | 7, 8 | 903 | |||
FQ216520.1 | - | 5, 6 | 642 | |||
FQ218274.1 | - | 7, 8 | 873 | |||
FQ219298.1 | - | 7, 8 | 875 | |||
FQ222930.1 | - | 7, 8 | 874 | |||
FQ223614.1 | - | 7, 8 | 873 | |||
FQ224956.1 | - | 6, 7 | 833 | |||
FQ226853.1 | - | 4, 5 | 546 | |||
FQ228703.1 | - | 7, 8 | 873 | |||
FQ228918.1 | - | 7, 8 | 875 | |||
FQ230995.1 | - | 7, 8 | 873 | |||
FQ231923.1 | - | 7, 8 | 875 | |||
FQ233868.1 | - | 7, 8 | 873 | |||
FQ234035.1 | - | 7, 8 | 873 | |||
FQ235034.1 | - | 7, 8 | 1025 |
Gene Symbol: Slamf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006250250.2 | XP_006250312.1 | 7 | 3144 | ||
GenBank mRNA | FQ230857.1 | - | 3 | 2078 |