Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1311300 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF196199.1 | - | 4 | 537 | ||
AF196209.1 | - | 4 | 277 | |||
AF196210.1 | - | 4 | 435 | |||
AF196211.1 | - | 4 | 464 | |||
AF196212.1 | - | 4 | 435 | |||
AF196213.1 | - | 3 | 427 | |||
AF196214.1 | - | 3 | 365 | |||
AF196215.1 | - | 3 | 438 | |||
AF196216.1 | - | 3 | 438 | |||
AF196217.1 | - | 474 | ||||
AF196218.1 | - | 481 | ||||
AF196219.1 | - | 474 | ||||
AF196220.1 | - | 474 | ||||
AF196221.1 | - | 478 | ||||
AF196222.1 | - | 477 | ||||
AF196223.1 | - | 393 | ||||
AF196224.1 | - | 492 | ||||
AF196225.1 | - | 506 | ||||
AF196226.1 | - | 514 | ||||
AF196227.1 | - | 368 | ||||
AF196228.1 | - | 478 | ||||
AF196229.1 | - | 5 | 410 | |||
AF196230.1 | - | 3 | 195 | |||
AF196231.1 | - | 4 | 506 | |||
AF196232.1 | - | 6 | 409 | |||
AF196234.1 | - | 4 | 491 | |||
AF196235.1 | - | 4 | 317 | |||
AF196236.1 | - | 4 | 517 | |||
AF196237.1 | - | 5 | 396 | |||
AF196238.1 | - | 6 | 420 | |||
AF196239.1 | - | 5 | 538 | |||
AF196240.1 | - | 5 | 424 | |||
AF196242.1 | - | 4 | 502 | |||
AF196243.1 | - | 5 | 398 | |||
AF196244.1 | - | 6 | 387 | |||
AF196245.1 | - | 5 | 476 | |||
AF196246.1 | - | 3 | 446 | |||
AF196247.1 | - | 6 | 387 | |||
AF196266.1 | - | 2 | 96 | |||
AF259788.1 | - | 3 | 317 | |||
AF259789.1 | - | 323 | ||||
AJ249228.1 | - | 4 | 182 | |||
BC099216.1 | - | 4 | 537 | 7384680 |