Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108925.2 | NP_001102395.1 | 7 | 534 | ||
GenBank mRNA | BC158732.1 | - | 7 | 534 | 9092746 |