Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nutm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108957.1 | NP_001102427.1 | 3 | 942 | ||
XM_006234708.2 | XP_006234770.1 | 3 | 938 |