Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flrt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126291.1 | NP_001119763.1 | 3 | 1971 | ||
GenBank mRNA | BC160843.1 | - | 3 | 1971 | 8361648 |