Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102550026 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763645.1 | XP_008761867.1 | 2 | 1636 |
Gene Symbol: Mob3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108970.1 | NP_001102440.1 | 3 | 984 | ||
XM_006238001.2 | XP_006238063.1 | 3 | 997 | |||
XM_008763630.1 | XP_008761852.1 | 3 | 1675 | |||
XM_008763631.1 | XP_008761853.1 | 5 | 1609 | |||
XM_008763632.1 | XP_008761854.1 | 4 | 1503 | |||
XM_008763633.1 | XP_008761855.1 | 3 | 1256 | |||
XM_008763634.1 | XP_008761856.1 | 3 | 1005 | |||
XM_008763635.1 | XP_008761857.1 | 5 | 2028 | |||
XM_008763636.1 | XP_008761858.1 | 3 | 1673 | |||
GenBank mRNA | FQ220583.1 | - | 2 | 475 | ||
FQ222144.1 | - | 3 | 942 |