Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257030.2 | XP_006257092.1 | 9 | 2150 | ||
XM_006257031.2 | XP_006257093.1 | 8 | 2430 | |||
XM_006257032.2 | XP_006257094.1 | 9 | 2497 | |||
XM_008773264.1 | XP_008771486.1 | 8 | 2082 | |||
XM_008773265.1 | XP_008771487.1 | 9 | 2531 | |||
XM_008773266.1 | XP_008771488.1 | 7 | 1932 |
Gene Symbol: LOC103694540 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773676.1 | XP_008771898.1 | 8 | 3110 | ||
XM_008773677.1 | XP_008771899.1 | 10 | 2251 | |||
XM_008773678.1 | XP_008771900.1 | 10 | 2245 | |||
XM_008773679.1 | XP_008771901.1 | 10 | 2226 | |||
XM_008773680.1 | XP_008771902.1 | 10 | 2183 |