Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109023.1 | NP_001102493.1 | 9 | 838 | ||
GenBank mRNA | FQ234501.1 | - | 9 | 847 |