Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iqsec3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207617.1 | NP_997500.1 | 3 | 1513 | ||
GenBank mRNA | AB057643.1 | - | 3 | 1513 |