Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1191 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000981.1 | NP_001000981.1 | 1 | 637 |
Gene Symbol: LOC100361194 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729873.3 | XP_002729919.2 | 1 | 637 |
Gene Symbol: Olr1192 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000859.1 | NP_001000859.1 | 1 | 637 |
Gene Symbol: Olr1193 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000594.1 | NP_001000594.1 | 1 | 637 |