Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010968.2 | NP_001010968.1 | 12 | 1921 | ||
GenBank mRNA | AY562420.1 | - | 12 | 1634 | ||
BC105860.1 | - | 12 | 1921 | 7456529 |