Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gucy1a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017090.2 | NP_058786.2 | 7, 8 | 1847 | ||
XM_006232518.2 | XP_006232580.1 | 8, 9 | 2439 | |||
XM_008761107.1 | XP_008759329.1 | 6, 7 | 2470 | |||
XM_008761108.1 | XP_008759330.1 | 7, 8 | 2213 | |||
GenBank mRNA | AB096020.1 | - | 6, 7 | 1710 | ||
BC085746.1 | - | 7, 8 | 1847 | 7190419 | ||
M57405.1 | - | 7, 8 | 2166 | |||
U60835.1 | - | 7, 8 | 2166 |