Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100789.1 | NP_001094259.1 | 35 | 6049 | ||
XM_006250672.2 | XP_006250734.1 | 35 | 6079 | |||
XM_006250673.2 | XP_006250735.1 | 34 | 5533 | |||
XM_006250674.2 | XP_006250736.1 | 32 | 5497 | |||
XM_008770051.1 | XP_008768273.1 | 32 | 5512 | |||
XM_008770052.1 | XP_008768274.1 | 35 | 6079 |
Gene Symbol: LOC100910240 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006250645.2 | XP_006250707.1 | 1063 |