Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gas2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127504.1 | NP_001120976.1 | 1 | 236 | ||
XM_006229270.2 | XP_006229332.1 | 2 | 701 | |||
XM_006229271.2 | XP_006229333.1 | 1 | 155 | |||
XM_006229273.2 | XP_006229335.1 | 2 | 493 | |||
XM_006229274.2 | XP_006229336.1 | 2 | 1020 | |||
XM_008759407.1 | XP_008757629.1 | 1 | 234 | |||
XM_008759408.1 | XP_008757630.1 | 1 | 155 |