Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109162.2 | NP_001102632.1 | 2 | 1487 | ||
XM_006230992.2 | XP_006231054.1 | 2 | 1405 | |||
GenBank mRNA | BC160822.1 | - | 2 | 1487 | 7380153 |