Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100910973 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | FM045317.1 | - | 2, 3 | 128 | ||
FM097063.1 | - | 2, 3 | 146 | |||
FM098643.1 | - | 2, 3 | 318 | |||
FM110991.1 | - | 2, 3 | 121 | |||
FQ212263.1 | - | 2, 3 | 142 | |||
FQ220597.1 | - | 2, 3 | 266 | |||
FQ230710.1 | - | 2, 3 | 142 | |||
FQ231880.1 | - | 2, 3 | 140 | |||
FQ234947.1 | - | 2, 3 | 138 |