Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024332.1 | NP_001019503.1 | 6 | 668 | ||
GenBank mRNA | BC081947.1 | - | 6 | 668 | 7121584 | |
FQ220718.1 | - | 6 | 660 |