Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d22b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025059.1 | NP_001020230.1 | 7, 8 | 907 | ||
GenBank mRNA | AY380821.1 | - | 7, 8 | 907 |