Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127572.1 | NP_001121044.1 | 6 | 410 | ||
GenBank mRNA | BC161949.1 | - | 6 | 410 | 9025530 |