Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnpc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100810.1 | NP_001094280.1 | 12, 13 | 1402 | ||
XM_006233211.2 | XP_006233273.1 | 12, 13 | 1780 | |||
XM_008761424.1 | XP_008759646.1 | 12, 13 | 1780 | |||
XM_008761425.1 | XP_008759647.1 | 12, 13 | 1781 | |||
GenBank mRNA | BC098742.1 | - | 10, 11 | 1080 | ||
BC166571.1 | - | 12, 13 | 1402 | 7320328 |