Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KIAA0319 SIRNA Assays ›
Gene Symbol:
KIAA0319
Gene Name:
KIAA0319
Gene Aliases:
DYLX2, DYX2, NMIG
Chromosome Location:
Chr.6: 24540131 - 24646211 on Build GRCh38
UniGene ID:
Hs.26441
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s19077
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KIAA0319
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001168374.1
NP_001161846.1
10
2103
NM_001168375.1
NP_001161847.1
9
1987
NM_001168376.1
NP_001161848.1
8
1827
NM_001168377.1
NP_001161849.1
9
2012
NM_001252328.1
NP_001239257.1
5
561
NM_014809.3
NP_055624.2
9
2012
XM_011515025.2
XP_011513327.1
9
1769
XM_011515026.2
XP_011513328.1
8
1730
XM_017011541.1
XP_016867030.1
10
2083
XM_017011542.1
XP_016867031.1
9
1730
XM_017011543.1
XP_016867032.1
8
1631
XM_017011544.1
XP_016867033.1
8
1704
XM_017011545.1
XP_016867034.1
9
1725
XM_017011546.1
XP_016867035.1
9
1730
XM_017011547.1
XP_016867036.1
9
1730
XM_017011548.1
XP_016867037.1
9
1803
XM_017011549.1
XP_016867038.1
9
1760
XM_017011550.1
XP_016867039.1
9
1730
XM_017011551.1
XP_016867040.1
9
1730
XM_017011552.1
XP_016867041.1
10
1687
XM_017011553.1
XP_016867042.1
10
1529
GenBank mRNA
AB002317.1
-
9
1998
AK295008.1
-
8
1564
AK296310.1
-
9
1694
AK296426.1
-
10
1838
AK307503.1
-
9
1697
AK310289.1
-
5
561
AL044313.1
-
1
91
BC140821.1
-
10
1843
9021689
BC144628.1
-
9
1871
9053159
BC152460.1
-
9
1998
8862596
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
neuron migration
negative regulation of dendrite development
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-849d5b7584-dvwjr:80/100.66.130.62:80.
git-commit: 0a9b6045e9144c2c4a21b698be73aa131cdbda4b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.32.0-2025.07.28-1.0