Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0319 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168374.1 | NP_001161846.1 | 4 | 1077 | ||
NM_001168375.1 | NP_001161847.1 | 3 | 961 | |||
NM_001168376.1 | NP_001161848.1 | 2 | 801 | |||
NM_001168377.1 | NP_001161849.1 | 3 | 986 | |||
NM_014809.3 | NP_055624.2 | 3 | 986 | |||
XM_011515025.2 | XP_011513327.1 | 3 | 743 | |||
XM_011515026.2 | XP_011513328.1 | 2 | 704 | |||
XM_017011541.1 | XP_016867030.1 | 4 | 1057 | |||
XM_017011542.1 | XP_016867031.1 | 3 | 704 | |||
XM_017011543.1 | XP_016867032.1 | 3 | 704 | |||
XM_017011544.1 | XP_016867033.1 | 3 | 777 | |||
XM_017011545.1 | XP_016867034.1 | 3 | 699 | |||
XM_017011546.1 | XP_016867035.1 | 3 | 704 | |||
XM_017011547.1 | XP_016867036.1 | 3 | 704 | |||
XM_017011548.1 | XP_016867037.1 | 3 | 777 | |||
XM_017011549.1 | XP_016867038.1 | 3 | 734 | |||
XM_017011550.1 | XP_016867039.1 | 3 | 704 | |||
XM_017011551.1 | XP_016867040.1 | 3 | 704 | |||
XM_017011552.1 | XP_016867041.1 | 4 | 661 | |||
XM_017011553.1 | XP_016867042.1 | 4 | 503 | |||
GenBank mRNA | AK295008.1 | - | 2 | 538 | ||
AK296310.1 | - | 3 | 668 | |||
AK296426.1 | - | 4 | 812 | |||
AK307503.1 | - | 3 | 671 | |||
BC140821.1 | - | 4 | 817 | 9021689 | ||
BC144628.1 | - | 3 | 845 | 9053159 |