Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CEP170 SIRNA Assays ›
          Gene Symbol:
          CEP170
          Gene Name:
          centrosomal protein 170
          Gene Aliases:
          FAM68A, KAB, KIAA0470
          Chromosome Location:
          Chr.1: 243124428 - 243255936 on Build GRCh38
          UniGene ID:
          Hs.533635
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s19086

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CEP170
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001042404.1 NP_001035863.1 5, 6 749
          NM_001042405.1 NP_001035864.1 5, 6 749
          NM_014812.2 NP_055627.2 5, 6 749
          XM_006711843.3 XP_006711906.1 5, 6 1272
          XM_011544334.2 XP_011542636.1 5, 6 1279
          XM_011544335.2 XP_011542637.1 5, 6 692
          XM_011544336.2 XP_011542638.1 5, 6 1598
          XM_011544337.2 XP_011542639.1 5, 6 1279
          XM_011544338.2 XP_011542640.1 5, 6 1279
          XM_011544339.2 XP_011542641.1 5, 6 1279
          XM_011544340.2 XP_011542642.1 5, 6 1229
          XM_011544341.2 XP_011542643.1 5, 6 1229
          XM_011544342.2 XP_011542644.1 5, 6 1229
          XM_011544343.2 XP_011542645.1 5, 6 1229
          XM_011544344.2 XP_011542646.1 5, 6 1279
          XM_017002932.1 XP_016858421.1 5, 6 1192
          XM_017002933.1 XP_016858422.1 5, 6 1049
          XM_017002934.1 XP_016858423.1 5, 6 692
          XM_017002935.1 XP_016858424.1 5, 6 1229
          XM_017002936.1 XP_016858425.1 5, 6 692
          XM_017002937.1 XP_016858426.1 5, 6 1229
          XM_017002938.1 XP_016858427.1 5, 6 1229
          XM_017002939.1 XP_016858428.1 5, 6 1049
          XM_017002940.1 XP_016858429.1 5, 6 692
          XM_017002941.1 XP_016858430.1 5, 6 1049
          XM_017002942.1 XP_016858431.1 5, 6 1229
          XM_017002943.1 XP_016858432.1 5, 6 1049
          XM_017002944.1 XP_016858433.1 5, 6 1272
          XM_017002945.1 XP_016858434.1 5, 6 1279
          XM_017002946.1 XP_016858435.1 5, 6 1229
          XM_017002947.1 XP_016858436.1 5, 6 1279
          XM_017002948.1 XP_016858437.1 5, 6 1272
          XM_017002949.1 XP_016858438.1 5, 6 1279
          XM_017002950.1 XP_016858439.1 5, 6 1229
          XM_017002951.1 XP_016858440.1 5, 6 1229
          GenBank mRNA AB007939.1 - 4, 5 394
          AB022657.1 - 4, 5 394
          AB022658.1 - 4, 5 394
          AB022659.1 - 4, 5 394
          AK296812.1 - 5, 6 398
          AK299552.1 - 5, 6 401
          BC050722.1 - 5, 6 664
          BC140794.1 - 4, 5 385 9021662
          BC146790.1 - 4, 5 394 9007254

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: XM_003751234, XM_006249682, XM_006249683, XM_006249681, XM_008769812, and others.

          Gene Ontology Categories:

          Process(es)

          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-867f8cf58d-sfwzd:80/100.66.130.62:80.
          git-commit: 7a7e1ef32845d1da800a461bcf18a29ea3e3afc0
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix