Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC688869 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145273.1 | NP_001138745.1 | 3 | 400 | ||
XM_006228795.1 | XP_006228857.1 | 4 | 508 | |||
GenBank mRNA | FQ217670.1 | - | 4 | 422 | ||
FQ217813.1 | - | 4 | 418 | |||
FQ218018.1 | - | 4 | 470 | |||
FQ221178.1 | - | 4 | 466 | |||
FQ222745.1 | - | 4 | 450 | |||
FQ223479.1 | - | 4 | 460 | |||
FQ223722.1 | - | 4 | 428 | |||
FQ223947.1 | - | 4 | 428 | |||
FQ224135.1 | - | 4 | 428 | |||
FQ226525.1 | - | 4 | 461 |