Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other UBAP2L SIRNA Assays ›
          Gene Symbol:
          UBAP2L
          Gene Name:
          ubiquitin associated protein 2 like
          Gene Aliases:
          NICE-4, NICE4
          Chromosome Location:
          Chr.1: 154220172 - 154271510 on Build GRCh38
          UniGene ID:
          Hs.490551
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s19176

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: UBAP2L
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001127320.2 NP_001120792.1 17 2105
          NM_001287815.1 NP_001274744.1 17 2138
          NM_001287816.1 NP_001274745.1 17 2192
          NM_014847.3 NP_055662.3 17 2159
          XM_005245658.3 XP_005245715.1 17 2192
          XM_005245667.3 XP_005245724.1 17 2192
          XM_005245668.3 XP_005245725.1 17 2192
          XM_005245669.3 XP_005245726.1 17 2192
          XM_005245670.3 XP_005245727.1 17 2192
          XM_005245672.3 XP_005245729.1 17 2084
          XM_005245673.3 XP_005245730.1 17 2192
          XM_005245674.1 XP_005245731.1 17 2084
          XM_011510215.2 XP_011508517.1 17 2192
          XM_011510216.1 XP_011508518.1 17 2084
          XM_017002972.1 XP_016858461.1 17 2217
          XM_017002973.1 XP_016858462.1 17 2084
          XM_017002974.1 XP_016858463.1 17 2115
          XM_017002975.1 XP_016858464.1 17 2192
          XM_017002976.1 XP_016858465.1 17 2094
          XM_017002977.1 XP_016858466.1 17 2084
          XM_017002978.1 XP_016858467.1 17 2115
          XM_017002979.1 XP_016858468.1 17 2076
          XM_017002980.1 XP_016858469.1 17 2115
          XM_017002981.1 XP_016858470.1 17 2076
          XM_017002982.1 XP_016858471.1 17 2084
          XM_017002983.1 XP_016858472.1 17 2115
          XM_017002984.1 XP_016858473.1 17 2115
          XM_017002985.1 XP_016858474.1 17 2217
          XM_017002986.1 XP_016858475.1 17 2084
          XM_017002987.1 XP_016858476.1 17 2115
          XM_017002988.1 XP_016858477.1 17 2115
          XM_017002989.1 XP_016858478.1 17 2115
          XM_017002990.1 XP_016858479.1 17 2076
          XM_017002991.1 XP_016858480.1 17 2115
          GenBank mRNA AK302662.1 - 17 2126
          AK302953.1 - 14 1853
          AK303533.1 - 17 2065
          BC003170.1 - 16 2041 2906083
          D63478.1 - 17 2098

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001024798, XM_006232706, XM_008761184, XM_008761182, XM_008761183, and others.
          This siRNA also targets the following transcripts in Mouse: NM_028475, NM_001165983, NM_001165987, NM_153489, NM_001165986, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          binding of sperm to zona pellucida
          hematopoietic stem cell homeostasis
          protein binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix